Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03936324_g1
          See other GM13497 GE Assays ›
          Gene Symbol:
          Gm13497
          Gene Name:
          predicted gene 13497
          Gene Aliases:
          A730015I17, OTTMUSG00000012425
          Chromosome Location:
          Chr.2: 51241831 - 51295945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03936324_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13497
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040636.1 2 - 3 545 97
          GenBank mRNA AK042686.1 - 2 - 3 546 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Rho family GTPase 3 Rnd3 2610017M01Rik,
          AI661404,
          Arhe,
          Rhoe
          -144.7kb Anti-sense Chr. 2 - 51130439 - 51149111 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700057H21 gene 1700057H21Rik 125.1kb Anti-sense Chr. 2 - 51421050 - 51437900 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000813.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148119 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04516478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04516460 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04516525 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102569 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04516460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01823522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02175566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02073273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02190060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01171136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01281221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01528765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01505190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01171135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01809494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01809511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01659480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01809500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01823523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02268298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01291322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01249622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01528757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01184711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01940325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01809508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01412797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01412791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01429080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01698219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01281219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02268300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02190057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01412798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01528772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01528768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01698204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01823526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01528767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21213065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02175563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01382966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21212605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200006726.mm.1 TC0200000813.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006726.mm.1 NR_040636.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006726.mm.1 NR_040636.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 502690 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101780632 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0