Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03932707_m1
          See other 6330410L21RIK GE Assays ›
          Gene Symbol:
          6330410L21Rik
          Gene Name:
          RIKEN cDNA 6330410L21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 129753758 - 129763882 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03932707_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6330410L21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040589.1 2 - 3 1340 119
          GenBank mRNA AK031828.1 - 2 - 3 1341 119

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream epidermal growth factor Egf AI790464 Anti-sense Chr. 3 - 129677569 - 129755322 on NCBI Build GRCm38
          Downstream leucine-rich repeat,
          immunoglobulin-like and transmembrane domains 3
          Lrit3 Gm421 30.3kb Anti-sense Chr. 3 - 129786929 - 129808884 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001311.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621903 AK007491.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621903 AK134732.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621903 AK167070.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621903 AK168269.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621903 BC054723.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621903 BC062876.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621903 NM_026932.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14298 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148857 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148857 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122097 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02583404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22521966 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300010581.mm.1 TC0300001311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010581.mm.1 NR_040589.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010581.mm.1 NR_040589.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010583.mm.1 TC0300001311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010583.mm.1 NR_040589.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010583.mm.1 NR_040589.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010584.mm.1 TC0300001311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010584.mm.1 NR_040589.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010584.mm.1 NR_040589.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010585.mm.1 TC0300001311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010585.mm.1 NR_040589.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010585.mm.1 NR_040589.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 746607 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103990731 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0