Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03931361_s1
          See other C430005N20RIK GE Assays ›
          Gene Symbol:
          C430005N20Rik
          Gene Name:
          RIKEN cDNA C430005N20 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03931361_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C430005N20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK021234.1 - 1 451 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY728941.1 - 1 449 Chr. 19 - 17632881 - 17633490 [-] on Build GRCm38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream riboflavin kinase Rfk 0610038L10Rik,
          AF031381,
          KOI-4
          -231.2kb Anti-sense Chr. 19 - 17394043 - 17401349 on NCBI Build GRCm38
          Downstream osteoclast stimulating factor 1 Ostf1 C78236,
          SH3P2,
          Sh3d3
          946.9kb Sense Chr. 19 - 18580364 - 18631813 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14351914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14146613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14267746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31987235 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100870348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0