Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300014917.mm.1 |
TC1300002282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014917.mm.1 |
NR_045965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014917.mm.1 |
NR_045965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014922.mm.1 |
TC1300002282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014922.mm.1 |
NR_045965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014922.mm.1 |
NR_045965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014920.mm.1 |
TC1300002282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014920.mm.1 |
NR_045965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014920.mm.1 |
NR_045965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014919.mm.1 |
TC1300002282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014919.mm.1 |
NR_045965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014919.mm.1 |
NR_045965.1 |
Refseq |