Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929153_s1
          See other FAM102B GE Assays ›
          Gene Symbol:
          Fam102b
          Gene Name:
          family with sequence similarity 102, member B
          Gene Aliases:
          1600010D10Rik, B430201A12Rik
          Chromosome Location:
          Chr.3: 108970997 - 109027607 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929153_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam102b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163567.1 NP_001157039.1 11 4962 103
          GenBank mRNA AK005412.1 - 1 145 103
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF616796.1 - 1 311 Chr. 3 - 108971220 - 108972047 [-] on Build GRCm38 103
          GenBankmRNA CK128340.1 - 1 114 Chr. 3 - 108971070 - 108971850 [-] on Build GRCm38 103
          GenBankmRNA BI690100.1 - 1 457 Chr. 3 - 108971391 - 108972196 [-] on Build GRCm38 103
          GenBankmRNA DT927380.1 - 1 125 Chr. 3 - 108971577 - 108971861 [-] on Build GRCm38 103
          GenBankmRNA CX243323.1 - 1 461 Chr. 3 - 108971401 - 108972197 [-] on Build GRCm38 103
          GenBankmRNA BE629111.1 - 1 271 Chr. 3 - 108971552 - 108972008 [-] on Build GRCm38 103
          GenBankmRNA BF118664.1 - 452 Chr. - - [-] on Build 103
          GenBankmRNA AA681812.1 - 1 247 Chr. 3 - 108971502 - 108971983 [-] on Build GRCm38 103
          GenBankmRNA AI573552.1 - 1 74 Chr. 3 - 108971419 - 108971810 [-] on Build GRCm38 103
          GenBankmRNA CB195831.1 - 1 141 Chr. 3 - 108971016 - 108971852 [-] on Build GRCm38 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream HEN1 methyltransferase homolog 1 (Arabidopsis) Henmt1 4921515J06Rik,
          Hen1,
          mHEN1
          -10.2kb Anti-sense Chr. 3 - 108938542 - 108960776 on NCBI Build GRCm38
          Downstream solute carrier family 25,
          member 54
          Slc25a54 4930443G12Rik,
          SCaMC-1like
          108.6kb Anti-sense Chr. 3 - 109080499 - 109120284 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002760.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300021743.mm.1 TC0300002760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300021743.mm.1 NM_001163567.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021743.mm.1 NM_001163567.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 488156 - -
          ABI Mouse Genome Survey Microarray 555515 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0