Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03808005_m1
          See other H13 GE Assays ›
          Gene Symbol:
          H13
          Gene Name:
          histocompatibility 13
          Gene Aliases:
          1200006O09Rik, 4930443L17Rik, 5031424B04Rik, AV020344, H-13, Hm13, PSL3, Spp
          Chromosome Location:
          Chr.2: 152669437 - 152708668 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03808005_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159551.1 NP_001153023.1 1 - 2 302 85
          NM_001159552.1 NP_001153024.1 1 - 2 302 85
          NM_001159553.1 NP_001153025.1 1 - 2 302 85
          NM_010376.4 NP_034506.1 1 - 2 302 85
          GenBank mRNA AF483214.1 - 1 - 2 269 85
          AF515662.1 - 1 - 2 181 85
          AJ345032.1 - 1 - 2 312 85
          AK004690.1 - 1 - 2 276 85
          AK007664.1 - 1 - 2 284 85
          AK012600.1 - 1 - 2 295 86
          AK019877.1 - 1 - 2 276 85
          AK028109.1 - 1 - 2 271 85
          AK039008.1 - 1 - 2 276 85
          AK049101.1 - 1 - 2 276 85
          AK050664.1 - 1 - 2 245 85
          AK051151.1 - 1 - 2 254 85
          AK080966.1 - 1 - 2 264 85
          AK159176.1 - 1 - 2 276 85
          AK169922.1 - 1 - 2 259 85
          AK171790.1 - 1 - 2 230 85
          BC056977.1 - 1 - 2 301 6818912 85
          CA321999.1 - 1 - 2 302 85
          DQ168449.1 - 1 - 2 318 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002295.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H13

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          membrane protein proteolysis
          aspartic-type endopeptidase activity
          peptidase activity
          hydrolase activity
          ubiquitin protein ligase binding
          aspartic endopeptidase activity, intramembrane cleaving
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 TC0200002295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_001159551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_001159551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_001159552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_001159552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_001159553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_001159553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_010376.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017939.mm.1 NM_010376.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 TC0200002295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_001159551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_001159551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_001159552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_001159552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_001159553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_001159553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_010376.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017940.mm.1 NM_010376.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 TC0200002295.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_001159551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_001159551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_001159552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_001159552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_001159553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_001159553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_010376.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017941.mm.1 NM_010376.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 915814 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0