Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pknox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027493.1 | 1 - 2 | 123 | 75 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NADH dehydrogenase (ubiquinone) flavoprotein 3 | Ndufv3 | 1500032D16Rik | -33.4kb | Sense | Chr. 17 - 31520115 - 31531326 on NCBI Build GRCm38 |
Downstream | cystathionine beta-synthase | Cbs |
AI047524, AI303044, HIP4 |
30.6kb | Anti-sense | Chr. 17 - 31612623 - 31637233 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() |