Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10473645 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10484864 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10473649 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10484877 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10484876 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10484878 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10473647 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10473644 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10484879 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10473646 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10484875 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10473643 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200010182.mm.1 |
TC0200001348.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010182.mm.1 |
NM_146263.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010182.mm.1 |
NM_146263.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034142.mm.1 |
TC0200004083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034142.mm.1 |
NM_146265.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034142.mm.1 |
NM_146265.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010177.mm.1 |
TC0200001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010177.mm.1 |
NM_001270423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010179.mm.1 |
TC0200001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010179.mm.1 |
NM_001270423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034141.mm.1 |
TC0200004083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034141.mm.1 |
NM_146265.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034141.mm.1 |
NM_146265.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200010178.mm.1 |
TC0200001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200010178.mm.1 |
NM_001270423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034140.mm.1 |
TC0200004083.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034140.mm.1 |
NM_146265.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034140.mm.1 |
NM_146265.2 |
Refseq |