Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02526285_s1
          See other XKR9 GE Assays ›
          Gene Symbol:
          Xkr9
          Gene Name:
          X Kell blood group precursor related family member 9 homolog
          Gene Aliases:
          Gm1620, XRG9
          Chromosome Location:
          Chr.1: 13668738 - 13701723 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02526285_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xkr9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011873.2 NP_001011873.1 4 1882 90
          GenBank mRNA AY534255.1 - 4 1882 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000071.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xkr9

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012261 NM_001011873.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012261 AY534255.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39934 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125686 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012261 NM_001011873.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012261 AY534255.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20042322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04018471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04018514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04018499 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04018469 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04018514 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00734599 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00734590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20042398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01029915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01112944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00820868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00734591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20042439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00820865 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10344925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10344927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10344926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10344928 - -
          Affymetrix Mouse Gene 1.1 ST Array 10344924 - -
          Affymetrix Mouse Clariom S Array PSR0100000634.mm.1 TC0100000071.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100000634.mm.1 NM_001011873.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000634.mm.1 NM_001011873.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229264 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3159558 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3080183 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3080183 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3159558 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0