Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02392633_s1
          See other EID2 GE Assays ›
          Gene Symbol:
          Eid2
          Gene Name:
          EP300 interacting inhibitor of differentiation 2
          Gene Aliases:
          Cri2, EID-2
          Chromosome Location:
          Chr.7: 28267881 - 28269168 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02392633_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eid2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198425.2 NP_940817.1 1 1170 96
          GenBank mRNA BC094513.1 - 1 1170 2648407 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eid2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transforming growth factor beta receptor complex assembly
          SMAD protein complex assembly
          multicellular organism development
          muscle organ development
          regulation of transforming growth factor beta receptor signaling pathway
          cell differentiation
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of cell population proliferation
          negative regulation of transcription, DNA-templated
          SMAD binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020882 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659762 NM_198425.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659762 AY251273.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659762 BC094513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659762 NM_198425.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659762 AY251273.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P659762 BC094513.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168857 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659762 NM_198425.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659762 AY251273.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659762 BC094513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659762 NM_198425.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659762 AY251273.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P659762 BC094513.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25020886 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168857 - -
          Whole Mouse Genome G4122A A_52_P659762 NM_198425.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659762 AY251273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659762 BC094513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659762 NM_198425.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659762 AY251273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P659762 BC094513.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168857 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06384075 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25020880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25020874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05946949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06054501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05950899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05910878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06636467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06567033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06123711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06248867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25020892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551486 - -
          Affymetrix HT MG-430 PM Array Plate 1454931_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551483 - -
          Mouse Genome 430 2.0 Array 1454931_at - -
          Mouse Genome 430 2.0 Array 1454931_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386655_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386655_at - -
          Mouse Expression Set 430 1454931_PM_at - -
          Mouse Expression Set 430 1454931_at - -
          Affymetrix HT Mouse Genome 430B Array 1454931_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454931_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3130176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223895 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223895 - -
          mouse-6 v1.1 (Illumina) scl0386655.1_75-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0386655.1_75-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223895 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223895 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0386655.1_75-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0