Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02029975_s1
          See other A930005H10RIK GE Assays ›
          Gene Symbol:
          A930005H10Rik
          Gene Name:
          RIKEN cDNA A930005H10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 115881578 - 115888130 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02029975_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A930005H10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015487.1 1 282 114
          NR_027894.1 1 211 114
          NR_027895.1 1 272 114
          GenBank mRNA AK007373.1 - 1 106 114
          AK020812.1 - 1 283 114
          AK081943.1 - 1 250 114
          AK137339.1 - 1 250 114
          BF019418.1 - 1 211 114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY234290.1 - 1 271 Chr. 3 - 115887737 - 115888120 [-] on Build GRCm38 114
          GenBankmRNA BF657927.1 - 1 249 Chr. 3 - 115881661 - 115888097 [-] on Build GRCm38 114
          GenBankmRNA W59576.1 - Chr. - - [-] on Build 113
          GenBankmRNA BB768725.1 - 1 179 Chr. 3 - 115881583 - 115888028 [-] on Build GRCm38 113
          GenBankmRNA BB628552.1 - 1 245 Chr. 3 - 115887486 - 115888093 [-] on Build GRCm38 114
          GenBankmRNA BB648213.1 - 1 246 Chr. 3 - 115887468 - 115888093 [-] on Build GRCm38 114
          GenBankmRNA BU559269.1 - 1 247 Chr. 3 - 115881612 - 115888092 [-] on Build GRCm38 114
          GenBankmRNA BY622656.1 - 1 70 Chr. 3 - 115881581 - 115887918 [-] on Build GRCm38 114
          GenBankmRNA AA016544.1 - 1 Chr. 3 - 115881733 - 115888006 [-] on Build GRCm38 113
          GenBankmRNA EL962384.1 - 1 255 Chr. 3 - 115881606 - 115888085 [-] on Build GRCm38 114
          GenBankmRNA BU604983.1 - 1 234 Chr. 3 - 115881578 - 115888083 [-] on Build GRCm38 114
          GenBankmRNA BY742122.1 - 1 283 Chr. 3 - 115881602 - 115888130 [-] on Build GRCm38 114

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream sphingosine-1-phosphate receptor 1 S1pr1 AI849002,
          Edg1,
          Lpb1,
          S1p,
          S1p1
          -172.7kb Sense Chr. 3 - 115710433 - 115715055 on NCBI Build GRCm38
          Downstream DPH5 homolog (S. cerevisiae) Dph5 2410012M04Rik,
          AU045680,
          C80186
          247.0 Anti-sense Chr. 3 - 115888165 - 115929323 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002799.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300022025.mm.1 TC0300002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300022025.mm.1 NR_015487.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022025.mm.1 NR_015487.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022025.mm.1 NR_027895.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022025.mm.1 NR_027895.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022018.mm.1 TC0300002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300022018.mm.1 NR_027894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022018.mm.1 NR_027894.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022024.mm.1 TC0300002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300022024.mm.1 NR_015487.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022024.mm.1 NR_015487.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022024.mm.1 NR_027894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022024.mm.1 NR_027894.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022024.mm.1 NR_027895.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022024.mm.1 NR_027895.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-98ffbb479-9qks8:80/100.66.134.79:80.
          git-commit: cc4aab6ff2cd04db3b50ee2b2ac7bb13b20eb648
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0