Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800003950.mm.1 |
TC1800000553.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800003950.mm.1 |
NR_045400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003950.mm.1 |
NR_045400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003949.mm.1 |
TC1800000553.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800003949.mm.1 |
NR_045400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003949.mm.1 |
NR_045400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003952.mm.1 |
TC1800000553.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800003952.mm.1 |
NR_045400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003952.mm.1 |
NR_045400.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003951.mm.1 |
TC1800000553.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800003951.mm.1 |
NR_045400.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003951.mm.1 |
NR_045400.1 |
Refseq |