Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01971942_g1
          See other MORF4L2 GE Assays ›
          Gene Symbol:
          Morf4l2
          Gene Name:
          mortality factor 4 like 2
          Gene Aliases:
          2410017O14Rik, Mrgx, Sid393p, mKIAA0026
          Chromosome Location:
          Chr.X: 136732942 - 136743686 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01971942_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Morf4l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001168225.1 NP_001161697.1 4 987 152
          NM_001168226.1 NP_001161698.1 4 920 152
          NM_001168227.1 NP_001161699.1 4 872 152
          NM_001168228.1 NP_001161700.1 4 871 152
          NM_001168229.1 NP_001161701.1 3 843 152
          NM_001168230.1 NP_001161702.1 3 781 152
          NM_019768.4 NP_062742.4 4 994 152
          GenBank mRNA AB025049.1 - 2 682 152
          AK075946.1 - 4 847 152
          AK088849.1 - 1 701 152
          AK089963.1 - 4 848 152
          AK129038.1 - 4 828 152
          AK131944.1 - 4 847 152
          AK145686.1 - 4 847 152
          AK153146.1 - 4 843 152
          AK159145.1 - 4 896 152
          AK159179.1 - 4 847 152
          AK160012.1 - 4 847 152
          AK161610.1 - 4 873 152
          AK164433.1 - 4 847 152
          AK165985.1 - 4 847 152
          AK166637.1 - 4 873 152
          BC075653.1 - 4 994 30539330 152
          BC088731.1 - 1 628 5340043 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003038.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Morf4l2

          Gene Ontology Categories:

          Function(s)

          DNA repair
          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          regulation of growth

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00019024.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019024.mm.1 NM_001168226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019024.mm.1 NM_001168226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019024.mm.1 NM_001168229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019024.mm.1 NM_001168229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019026.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019026.mm.1 NM_019768.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019026.mm.1 NM_019768.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168228.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168228.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168230.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_001168230.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_019768.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019009.mm.1 NM_019768.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168228.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168228.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168230.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_001168230.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_019768.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019010.mm.1 NM_019768.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019029.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019029.mm.1 NM_001168227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019029.mm.1 NM_001168227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168228.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168228.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168230.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_001168230.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_019768.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019008.mm.1 NM_019768.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019023.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019023.mm.1 NM_001168226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019023.mm.1 NM_001168226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019023.mm.1 NM_001168229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019023.mm.1 NM_001168229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019028.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019028.mm.1 NM_001168227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019028.mm.1 NM_001168227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168225.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168225.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168226.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168226.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168228.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168228.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168230.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_001168230.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_019768.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019007.mm.1 NM_019768.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019030.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019030.mm.1 NM_001168227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019030.mm.1 NM_001168227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019031.mm.1 TC0X00003038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00019031.mm.1 NM_001168228.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019031.mm.1 NM_001168228.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019031.mm.1 NM_001168230.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00019031.mm.1 NM_001168230.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897671 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0