Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm9837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076795.1 | - | 1 | 674 | 124 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | shroom family member 1 | Shroom1 |
Apx, Shrm1 |
-2.0kb | Anti-sense | Chr. 11 - 53456709 - 53467766 on NCBI Build GRCm38 |
Downstream | sosondowah ankyrin repeat domain family member A | Sowaha |
A830006N08Rik, Ankrd43 |
6.1kb | Sense | Chr. 11 - 53476578 - 53480195 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |