Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01960808_u1
          See other SPINK7 GE Assays ›
          Gene Symbol:
          Spink7
          Gene Name:
          serine peptidase inhibitor, Kazal type 7 (putative)
          Gene Aliases:
          EG408198, Ecg2
          Chromosome Location:
          Chr.18: 62592413 - 62596264 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01960808_u1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spink7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001803.2 NP_001001803.2 1 48 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001443.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spink7

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of peptidase activity
          molecular_function
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051983 NM_001001803.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135239 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051983 NM_001001803.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31764575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10459319 - -
          Affymetrix Mouse Clariom S Array PSR1800010608.mm.1 TC1800001443.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010608.mm.1 NM_001001803.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010608.mm.1 NM_001001803.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010605.mm.1 TC1800001443.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010605.mm.1 NM_001001803.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010605.mm.1 NM_001001803.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010606.mm.1 TC1800001443.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010606.mm.1 NM_001001803.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010606.mm.1 NM_001001803.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010609.mm.1 TC1800001443.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800010609.mm.1 NM_001001803.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800010609.mm.1 NM_001001803.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161810 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161810 - -
          mouse-6 v1.1 (Illumina) GI_49227051-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161810 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161810 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0