Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01740552_g1
          See other RDH18-PS GE Assays ›
          Gene Symbol:
          Rdh18-ps
          Gene Name:
          retinol dehydrogenase 18, pseudogene
          Gene Aliases:
          Rdh18
          Chromosome Location:
          Chr.10: 127824216 - 127846565 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01740552_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rdh18-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037604.1 3 - 4 808 148
          GenBank mRNA AY053573.1 - 3 - 4 740 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003222.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2095859 NR_037604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295012 AY053573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295012 NR_037604.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P27538239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27538234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27538237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27538236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10367053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10367051 - -
          Affymetrix HT MG-430 PM Array Plate 1445728_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10367050 - -
          Mouse Genome 430 2.0 Array 1445728_at - -
          Mouse Genome 430 2.0 Array 1445728_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000011359.mm.1 TC1000003222.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011359.mm.1 NR_037604.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011357.mm.1 TC1000003222.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011357.mm.1 NR_037604.1 Refseq
          Mouse Expression Set 430 1445728_at - -
          Mouse Expression Set 430 1445728_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445728_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445728_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213579 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213579 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0