Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01716308_m1
          See other 2310040G24RIK GE Assays ›
          Gene Symbol:
          2310040G24Rik
          Gene Name:
          RIKEN cDNA 2310040G24 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 86483376 - 86488227 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01716308_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310040G24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040292.1 1 - 2 431 72
          GenBank mRNA AK009724.1 - 1 - 2 431 72
          BC070414.1 - 1 - 2 153 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002637.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159730 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119350 AK009724.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119350 BC070414.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01375580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05317041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05499922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05698714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05501780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05268528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24610359 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1443575_PM_at - -
          Mouse Genome 430 2.0 Array 1443575_PM_at - -
          Mouse Genome 430 2.0 Array 1443575_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381792_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381792_at - -
          Affymetrix Mouse Clariom S Array PSR0600020476.mm.1 TC0600002637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020476.mm.1 NR_040292.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020476.mm.1 NR_040292.1 Refseq
          Mouse Expression Set 430 1443575_PM_at - -
          Mouse Expression Set 430 1443575_at - -
          Murine Genome U74v2 Set 167584_at - -
          Murine Genome U74v2 Set 167584_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443575_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443575_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 784425 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0