Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01716220_m1
          See other 4930420G21RIK GE Assays ›
          Gene Symbol:
          4930420G21Rik
          Gene Name:
          RIKEN cDNA 4930420G21 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01716220_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930420G21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK054212.1 - 1 - 2 141 113

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345388 U39074.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345388 U39075.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345388 U39076.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345388 U39077.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124697 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137730 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08675540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08675572 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137730 - -
          Whole Mouse Genome G4122A A_52_P102300 AK019579.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P102300 AK054212.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121056 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08675572 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431621_PM_at - -
          Mouse Genome 430 2.0 Array 1431621_at - -
          Mouse Genome 430 2.0 Array 1431621_PM_at - -
          Mouse Expression Set 430 1431621_PM_at - -
          Mouse Expression Set 430 1431621_at - -
          Affymetrix HT Mouse Genome 430B Array 1431621_at - -
          Affymetrix HT Mouse Genome 430B Array 1431621_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 382011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0