Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01703075_gH
          See other MND1 GE Assays ›
          Gene Symbol:
          Mnd1 Mnd1-ps
          Gene Name:
          meiotic nuclear divisions 1 homolog (S. cerevisiae)
          Mnd1 retrotransposed pseudogene
          Gene Aliases:
          2610034E18Rik, AI591601, Gaj
          Gm3833
          Chromosome Location:
          Chr.3: 84087933 - 84156861 on Build GRCm38
          Chromosome Location:
          Chr.14: 9884095 - 9886868 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01703075_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mnd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029797.3 NP_084073.2 4 - 5 399 116
          GenBank mRNA AK011664.1 - 4 - 5 309 116
          BC027741.1 - 4 - 5 326 5035682 116
          BY710901.1 - 4 - 5 309 116
          Gene Symbol: Mnd1-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030680.1 1 2199 116
          GenBank mRNA AK156325.1 - 1 2099 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002241.mm.1, TC1400000088.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mnd1
          Click here to find siRNAs for Gene Mnd1-ps

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA recombination
          meiotic cell cycle
          DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10499035 - -
          Affymetrix Mouse Clariom S Array PSR1400000753.mm.1 TC1400000088.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400000753.mm.1 NR_030680.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016848.mm.1 TC0300002241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016848.mm.1 NM_029797.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016848.mm.1 NM_029797.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016849.mm.1 TC0300002241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016849.mm.1 NM_029797.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016849.mm.1 NM_029797.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000752.mm.1 TC1400000088.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400000752.mm.1 NR_030680.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000754.mm.1 TC1400000088.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400000754.mm.1 NR_030680.1 Refseq
          Murine Genome U74v2 Set 165637_i_at - -
          Murine Genome U74v2 Set 165637_PM_i_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0