Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01617623_g1
          See other TRMT112 GE Assays ›
          Gene Symbol:
          Trmt112
          Gene Name:
          tRNA methyltransferase 11-2
          Gene Aliases:
          0610038D11Rik, Trm112p
          Chromosome Location:
          Chr.19: 6909698 - 6911028 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01617623_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trmt112
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166370.1 NP_001159842.1 3 - 4 752 123
          NM_026306.3 NP_080582.3 4 - 5 654 123
          GenBank mRNA AK002791.1 - 3 - 4 299 123
          AK003292.1 - 3 - 4 123
          BC016191.1 - 3 - 4 674 4483787 123
          BC087959.1 - 4 - 5 654 6766768 123
          BE951374.1 - 1 - 2 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000124.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trmt112

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-glutamine methylation
          protein methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936615 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936619 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936612 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936616 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936617 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936613 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987600 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127131 BC016191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127131 BC019418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127131 BC087959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127131 BE951374.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127131 NM_026306.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127118 BC016191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127118 BC087959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127118 NM_026306.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127118 DV654053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127118 NM_001166370.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168444 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987600 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127131 BC016191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127131 BC019418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127131 BC087959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127131 BE951374.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127131 NM_026306.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127118 BC016191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127118 BC087959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127118 NM_026306.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127118 DV654053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127118 NM_001166370.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936620 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168444 - -
          Whole Mouse Genome G4122A A_51_P203022 BC016191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203022 BC019418.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203022 BC087959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203022 BE951374.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203022 NM_026306.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128099 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127118 BC016191.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127118 BC087959.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127118 NM_026306.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127118 DV654053.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127118 NM_001166370.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14248031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14248005 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14248032 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14248015 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14247992 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936615 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936612 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936617 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936616 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936619 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14248031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531285 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461014 - -
          Affymetrix HT MG-430 PM Array Plate 1460375_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10531284 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510462 - -
          Affymetrix Mouse Gene 1.1 ST Array 10465474 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461012 - -
          Mouse Genome 430 2.0 Array 1425299_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425299_s_at - -
          Mouse Genome 430 2.0 Array 1460375_PM_at - -
          Mouse Genome 430 2.0 Array 1460375_at - -
          Affymetrix Mouse Clariom S Array PSR1900001689.mm.1 TC1900000124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001689.mm.1 NM_001166370.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001689.mm.1 NM_001166370.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001689.mm.1 NM_026306.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001689.mm.1 NM_026306.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001690.mm.1 TC1900000124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001690.mm.1 NM_001166370.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001690.mm.1 NM_001166370.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001690.mm.1 NM_026306.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001690.mm.1 NM_026306.3 Refseq
          Mouse Expression Set 430 1460375_PM_at - -
          Mouse Expression Set 430 1425299_s_at - -
          Mouse Expression Set 430 1460375_at - -
          Mouse Expression Set 430 1425299_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1460375_PM_at - -
          Mouse Genome 430A 2.0 Array 1425299_s_at - -
          Mouse Genome 430A 2.0 Array 1460375_at - -
          Mouse Genome 430A 2.0 Array 1425299_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1460375_at - -
          Affymetrix HT Mouse Genome 430A Array 1460375_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 778542 - -
          ABI Mouse Genome Survey Microarray 896600 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101400333 - -
          Illumina mouse-6 v1.0 expression beadchip 101400333 - -
          Illumina mouse-6 v1.0 expression beadchip 4120519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737319 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2470615 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737319 - -
          mouse-6 v1.1 (Illumina) scl067674.3_12-S - -
          mouse-6 v1.1 (Illumina) scl53555.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 4120519 - -
          Illumina Sentrix mouse-6.v1 bead array 101400333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067674.3_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120519 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101400333 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2470615 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737319 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067674.3_12-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0