Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335754_m1
          See other GM16499 GE Assays ›
          Gene Symbol:
          Gm16499
          Gene Name:
          predicted gene 16499
          Gene Aliases:
          ENSMUSG00000068503
          Chromosome Location:
          Chr.6: 53844899 - 53867140 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335754_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm16499
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK039090.1 - 659 95
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CO803841.1 - 2 - 3 177 Chr. 6 - 53858528 - 53867142 [+] on Build GRCm38 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TLR4 interactor with leucine-rich repeats Tril 1200009O22Rik,
          AI316813,
          mKIAA0644
          -38.0kb Anti-sense Chr. 6 - 53815468 - 53820825 on NCBI Build GRCm38
          Downstream carboxypeptidase,
          vitellogenic-like
          Cpvl 4933436L16Rik,
          HVLP
          6.1kb Anti-sense Chr. 6 - 53873279 - 53978673 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098776 BC006619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098776 BC043099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098776 BC059224.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098776 BY291576.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098776 NM_025817.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676480 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 497995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863116 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54607 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163759 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100885 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098776 BC006619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098776 BC043099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098776 BC059224.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098776 BY291576.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098776 NM_025817.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01332158 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05903392 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05903401 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05903379 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100885 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01332137 - -
          Whole Mouse Genome G4122A A_51_P465758 BC006619.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126794 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01332137 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163759 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05763890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05696389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05151814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05413546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05204284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05819798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05854635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05223062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05237132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05678405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05541116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05373314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05541109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24459586 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1427643_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440733_PM_at - -
          Mouse Genome 430 2.0 Array 1440733_PM_at - -
          Mouse Genome 430 2.0 Array 1427643_PM_at - -
          Mouse Genome 430 2.0 Array 1427643_at - -
          Mouse Genome 430 2.0 Array 1440733_at - -
          Mouse Expression Set 430 1427643_at - -
          Mouse Expression Set 430 1427643_PM_at - -
          Mouse Expression Set 430 1440733_at - -
          Mouse Expression Set 430 1440733_PM_at - -
          Mouse Genome 430A 2.0 Array 1427643_at - -
          Mouse Genome 430A 2.0 Array 1427643_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427643_at - -
          Affymetrix HT Mouse Genome 430A Array 1427643_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440733_at - -
          Affymetrix HT Mouse Genome 430B Array 1440733_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0