Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335293_g1
          See other NRON GE Assays ›
          Gene Symbol:
          Nron
          Gene Name:
          non-protein coding RNA, repressor of NFAT
          Gene Aliases:
          A630071L07Rik, Gm24353
          Chromosome Location:
          Chr.2: 33805816 - 33813591 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335293_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nron
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045729.1 2 - 3 759 88
          GenBank mRNA AK037308.1 - 2 - 3 748 88
          AK042215.1 - 2 - 3 448 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 9430024E24 gene 9430024E24Rik -90.8kb Anti-sense Chr. 2 - 33714606 - 33718899 on NCBI Build GRCm38
          Downstream predicted gene 13403 Gm13403 OTTMUSG00000011915 122.5kb Anti-sense Chr. 2 - 33936123 - 33942145 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000609.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          regulation of cytokine production
          negative regulation of dephosphorylation
          regulation of calcineurin-NFAT signaling cascade

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2207335 AK042215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2207335 AK042215.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106988 - -
          Agilent Mouse cDNA Microarray (G4104A) 1281494 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1457187_PM_at - -
          Mouse Genome 430 2.0 Array 1457187_at - -
          Mouse Genome 430 2.0 Array 1457187_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200005604.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005604.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005604.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005599.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005599.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005599.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005595.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005595.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005595.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005600.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005600.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005600.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005603.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005603.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005603.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005597.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005597.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005597.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005598.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005598.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005598.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005602.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005602.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005602.mm.1 NR_045729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005596.mm.1 TC0200000609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005596.mm.1 NR_045729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005596.mm.1 NR_045729.1 Refseq
          Mouse Expression Set 430 1457187_at - -
          Mouse Expression Set 430 1457187_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457187_at - -
          Affymetrix HT Mouse Genome 430B Array 1457187_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 303257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0