Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333459_g1
          See other PTRH1 GE Assays ›
          Gene Symbol:
          Ptrh1
          Gene Name:
          peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae)
          Gene Aliases:
          2210013M04Rik, AW046744
          Chromosome Location:
          Chr.2: 32775738 - 32777593 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333459_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptrh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178595.3 NP_848710.1 4 - 5 439 104
          GenBank mRNA AK075833.1 - 4 - 5 440 104
          BC098218.1 - 4 - 5 479 3466952 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000593.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptrh1

          Gene Ontology Categories:

          Process(es)

          RNA binding
          aminoacyl-tRNA hydrolase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21117415 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 NM_178595.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 AK075833.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 BC098218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 BC118617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 BC121825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 NM_178595.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 AK075833.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 BC098218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 BC118617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P605812 BC121825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042462 AK132137.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358890 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162976 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148016 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 NM_178595.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 AK075833.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 BC098218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 BC118617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 BC121825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 NM_178595.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 AK075833.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 BC098218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 BC118617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P605812 BC121825.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21117453 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148016 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162976 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04477459 - -
          Whole Mouse Genome G4122A A_52_P605812 NM_178595.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 AK075833.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 BC098218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 BC118617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 BC121825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 NM_178595.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 AK075833.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 BC098218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 BC118617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605812 BC121825.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122453 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162976 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01713982 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01713997 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21117415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21117482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21117425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01950909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01713984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21117417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21117421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21117419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21117424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21117426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01713982 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471529 - -
          Affymetrix HT MG-430 PM Array Plate 1436336_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10471525 - -
          Affymetrix Mouse Gene 1.1 ST Array 10481700 - -
          Mouse Genome 430 2.0 Array 1436336_at - -
          Mouse Genome 430 2.0 Array 1436336_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329384_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329384_at - -
          Affymetrix Mouse Clariom S Array PSR0200005481.mm.1 TC0200000593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005481.mm.1 NM_178595.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005481.mm.1 NM_178595.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005480.mm.1 TC0200000593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005480.mm.1 NM_178595.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005480.mm.1 NM_178595.3 Refseq
          Mouse Expression Set 430 1436336_PM_at - -
          Mouse Expression Set 430 1436336_at - -
          Murine Genome U74v2 Set 103601_at - -
          Murine Genome U74v2 Set 103601_PM_at - -
          Mouse Genome 430A 2.0 Array 1436336_PM_at - -
          Mouse Genome 430A 2.0 Array 1436336_at - -
          Affymetrix HT Mouse Genome 430A Array 1436336_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436336_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 544530 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589615 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2589615 - -
          mouse-6 v1.1 (Illumina) scl21050.3.17_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21050.3.17_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2812645 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2589615 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2812645 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589615 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5db945b66b-v5hlb:80/100.66.134.185:80.
          git-commit: 84360441031e17f3391aad9879d55c303cd12901
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0