Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01332053_m1
          See other 4930408O17RIK GE Assays ›
          Gene Symbol:
          4930408O17Rik
          Gene Name:
          RIKEN cDNA 4930408O17 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01332053_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930408O17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015114.1 - 1 - 2 164 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714660.1 - 1 - 2 164 Chr. 12 - 104865330 - 104871525 [+] on Build GRCm38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream calmin Clmn 9330188N17Rik,
          AI428889,
          AI788815,
          mKIAA1188
          -254.0 Anti-sense Chr. 12 - 104763114 - 104865076 on NCBI Build GRCm38
          Downstream spectrin repeat containing,
          nuclear envelope family member 3
          Syne3 nesprin-3 58.3kb Anti-sense Chr. 12 - 104929830 - 105010098 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P410516 AK015114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P410516 AK015114.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07735728 - -
          Whole Mouse Genome G4122A A_51_P410516 AK015114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P410516 AK015114.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137185 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10410579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28706074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28706075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28706073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28706082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09804640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10166993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10166982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10166991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10166981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28706078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432541_PM_at - -
          Mouse Genome 430 2.0 Array 1432541_PM_at - -
          Mouse Genome 430 2.0 Array 1432541_at - -
          Mouse Expression Set 430 1432541_at - -
          Mouse Expression Set 430 1432541_PM_at - -
          Murine Genome U74v2 Set 169972_PM_at - -
          Murine Genome U74v2 Set 169972_at - -
          Affymetrix HT Mouse Genome 430B Array 1432541_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432541_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 559665 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0