Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331000_m1
          See other ZFP862-PS GE Assays ›
          Gene Symbol:
          Zfp862-ps
          Gene Name:
          zinc finger protein 862, pseudogene
          Gene Aliases:
          4732460K03Rik, AI448302, Zfp862, mKIAA0543
          Chromosome Location:
          Chr.6: 48504339 - 48534832 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331000_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp862-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015597.1 3 - 4 1024 70
          GenBank mRNA AK129160.1 - 3 - 4 946 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000554.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878901 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 162689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90608 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178212 - -
          Whole Mouse Genome G4122A A_52_P555629 BC037484.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555629 BC059055.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555629 BC068148.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555629 AK129160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555629 AK028840.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127827 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05136639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05136634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05136638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05375594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05375593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05155962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05128939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05585791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05155964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05346988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05025303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05375597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05170811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24427679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05375595 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1434591_PM_at - -
          Mouse Genome 430 2.0 Array 1434591_PM_at - -
          Mouse Genome 430 2.0 Array 1434591_at - -
          Affymetrix Mouse Clariom S Array PSR0600004243.mm.1 TC0600000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004243.mm.1 NR_015597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004243.mm.1 NR_015597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004243.mm.1 NR_027808.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004243.mm.1 NR_027808.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004245.mm.1 TC0600000554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004245.mm.1 NR_015597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004245.mm.1 NR_015597.1 Refseq
          Mouse Expression Set 430 1434591_at - -
          Mouse Expression Set 430 1434591_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434591_at - -
          Affymetrix HT Mouse Genome 430B Array 1434591_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2461582 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0