Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01324885_m1
          See other MVB12B GE Assays ›
          Gene Symbol:
          Mvb12b
          Gene Name:
          multivesicular body subunit 12B
          Gene Aliases:
          2610200O14Rik, 2610528K11Rik, AI414895, Fam125b
          Chromosome Location:
          Chr.2: 33729953 - 33887946 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01324885_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mvb12b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175184.4 NP_780393.2 2 - 3 754 62
          GenBank mRNA AK087821.1 - 2 - 3 272 62
          AK131113.1 - 2 - 3 272 62
          AK135990.1 - 2 - 3 188 62
          AK154267.1 - 2 - 3 248 62
          BC049129.1 - 2 - 3 267 6825067 62
          BC059907.1 - 2 - 3 754 6824889 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003332.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mvb12b

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          protein transport
          virus maturation
          regulation of epidermal growth factor receptor signaling pathway
          positive regulation of viral process
          lipid binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200028515.mm.1 TC0200003332.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028515.mm.1 NM_175184.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028515.mm.1 NM_175184.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028519.mm.1 TC0200003332.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028519.mm.1 NM_175184.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028519.mm.1 NM_175184.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028521.mm.1 TC0200003332.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028521.mm.1 NM_175184.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028521.mm.1 NM_175184.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028517.mm.1 TC0200003332.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028517.mm.1 NM_175184.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028517.mm.1 NM_175184.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 798189 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0