Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01322194_m1
          See other ADAMDEC1 GE Assays ›
          Gene Symbol:
          Adamdec1
          Gene Name:
          ADAM-like, decysin 1
          Gene Aliases:
          2210414L24Rik, AI574231, Dcsn
          Chromosome Location:
          Chr.14: 68563387 - 68582072 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01322194_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adamdec1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021475.2 NP_067450.1 1 - 2 111 126
          GenBank mRNA AJ242912.1 - 1 - 2 111 126
          AK156586.1 - 1 - 2 126 126
          BC026542.1 - 1 - 2 134 126
          BC046324.1 - 1 - 2 102 6394673 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002302.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adamdec1

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          metalloendopeptidase activity
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400017828.mm.1 TC1400002302.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017828.mm.1 NM_021475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017828.mm.1 NM_021475.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017830.mm.1 TC1400002302.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017830.mm.1 NM_021475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017830.mm.1 NM_021475.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017827.mm.1 TC1400002302.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017827.mm.1 NM_021475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017827.mm.1 NM_021475.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017826.mm.1 TC1400002302.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017826.mm.1 NM_021475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017826.mm.1 NM_021475.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017829.mm.1 TC1400002302.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017829.mm.1 NM_021475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017829.mm.1 NM_021475.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0