Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100014558.mm.1 |
TC0100001745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100014558.mm.1 |
NM_173378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100014558.mm.1 |
NM_173378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100014556.mm.1 |
TC0100001745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100014556.mm.1 |
NM_173378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100014556.mm.1 |
NM_173378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100014557.mm.1 |
TC0100001745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100014557.mm.1 |
NM_173378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100014557.mm.1 |
NM_173378.2 |
Refseq |