Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01316755_m1
          See other RAB10OS GE Assays ›
          Gene Symbol:
          Rab10os
          Gene Name:
          RAB10, member RAS oncogene family, opposite strand
          Gene Aliases:
          1700012B15Rik, 1810036A22Rik
          Chromosome Location:
          Chr.12: 3235791 - 3250374 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01316755_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab10os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015551.1 6 - 7 565 86
          GenBank mRNA AK007702.1 - 6 - 7 566 86
          BC080728.1 - 6 - 7 2362 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000005.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322123 BC080728.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322123 AK007702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322123 NR_015551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322123 BC080728.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322123 AK007702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P322123 NR_015551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136813 BC099424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136813 BC062897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136813 AK005891.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136813 AK171768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136813 BC038315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095354 AK005891.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2095354 AK171768.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07520697 - -
          Whole Mouse Genome G4122A A_52_P361338 AK005891.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P361338 BC060685.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28194118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10394157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394155 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394146 - -
          Affymetrix HT MG-430 PM Array Plate 1453294_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431818_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10394144 - -
          Mouse Genome 430 2.0 Array 1453294_PM_at - -
          Mouse Genome 430 2.0 Array 1431818_at - -
          Mouse Genome 430 2.0 Array 1431818_PM_at - -
          Mouse Genome 430 2.0 Array 1453294_at - -
          Affymetrix Mouse Clariom S Array PSR1200000047.mm.1 TC1200000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000047.mm.1 NR_015551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000047.mm.1 NR_015551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000049.mm.1 TC1200000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000049.mm.1 NR_015551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000049.mm.1 NR_015551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000046.mm.1 TC1200000005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200000046.mm.1 NR_015551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200000046.mm.1 NR_015551.1 Refseq
          Mouse Expression Set 430 1431818_at - -
          Mouse Expression Set 430 1431818_PM_at - -
          Mouse Expression Set 430 1453294_at - -
          Mouse Expression Set 430 1453294_PM_at - -
          Murine Genome U74v2 Set 95924_at - -
          Murine Genome U74v2 Set 95924_PM_at - -
          Murine Genome U74v2 Set 171085_at - -
          Murine Genome U74v2 Set 171085_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453294_at - -
          Affymetrix HT Mouse Genome 430B Array 1431818_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431818_at - -
          Affymetrix HT Mouse Genome 430B Array 1453294_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 686931 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 580605 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0