Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01316546_g1
          See other KRIT1 GE Assays ›
          Gene Symbol:
          Krit1
          Gene Name:
          KRIT1, ankyrin repeat containing
          Gene Aliases:
          2010007K12Rik, A630036P20Rik, AA432855, AI450393, AI643869, BB155247, BB235701, Ccm1
          Chromosome Location:
          Chr.5: 3803165 - 3844515 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01316546_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krit1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170552.1 NP_001164023.1 14 - 15 2179 62
          NM_030675.3 NP_109600.2 14 - 15 2133 62
          NR_033173.1 15 - 16 2380 62
          GenBank mRNA AF306509.1 - 14 - 15 1828 62
          AF310134.1 - 14 - 15 2109 62
          AK039264.1 - 2010 62
          AK041574.1 - 3 - 4 265 62
          AK085107.1 - 13 - 14 3554 62
          AK144667.1 - 14 - 15 2126 62
          AY328895.1 - 13 - 14 1815 62
          AY464945.1 - 13 - 14 1815 62
          BC016087.1 - 14 - 15 2083 62
          BC044820.1 - 14 - 15 2022 62
          BC054819.1 - 15 - 16 2372 62
          BC085252.1 - 14 - 15 1939 62
          BC141042.1 - 14 - 15 1977 9055654 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000019.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krit1

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of endothelial cell proliferation
          negative regulation of endothelial cell migration
          negative regulation of angiogenesis
          positive regulation of protein binding
          cell redox homeostasis
          regulation of catalytic activity
          regulation of establishment of cell polarity
          negative regulation of endothelial cell apoptotic process
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          microtubule binding
          GTPase regulator activity
          protein-containing complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500000190.mm.1 TC0500000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000190.mm.1 NM_001170552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000190.mm.1 NM_001170552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000190.mm.1 NM_030675.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000190.mm.1 NM_030675.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000190.mm.1 NR_033173.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000190.mm.1 NR_033173.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000191.mm.1 TC0500000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000191.mm.1 NM_030675.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000191.mm.1 NM_030675.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000191.mm.1 NR_033173.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000191.mm.1 NR_033173.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000192.mm.1 TC0500000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000192.mm.1 NM_001170552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000192.mm.1 NM_001170552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000192.mm.1 NM_030675.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000192.mm.1 NM_030675.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000192.mm.1 NR_033173.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000192.mm.1 NR_033173.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000193.mm.1 TC0500000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000193.mm.1 NM_001170552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000193.mm.1 NM_001170552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000193.mm.1 NM_030675.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000193.mm.1 NM_030675.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000193.mm.1 NR_033173.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000193.mm.1 NR_033173.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000194.mm.1 TC0500000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000194.mm.1 NM_001170552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000194.mm.1 NM_001170552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000194.mm.1 NM_030675.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000194.mm.1 NM_030675.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000194.mm.1 NR_033173.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000194.mm.1 NR_033173.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000189.mm.1 TC0500000019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000189.mm.1 NM_001170552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000189.mm.1 NM_001170552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000189.mm.1 NM_030675.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000189.mm.1 NM_030675.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000189.mm.1 NR_033173.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000189.mm.1 NR_033173.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-55c5c7589b-pgs97:80/100.66.130.250:80.
          git-commit: fefcbad4eb9baa7f43a2c44c49530f643471c973
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0