Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700031089.mm.1 |
TC0700003657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031089.mm.1 |
NM_010176.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031089.mm.1 |
NM_010176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031090.mm.1 |
TC0700003657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031090.mm.1 |
NM_010176.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031090.mm.1 |
NM_010176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031088.mm.1 |
TC0700003657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031088.mm.1 |
NM_010176.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031088.mm.1 |
NM_010176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031087.mm.1 |
TC0700003657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700031087.mm.1 |
NM_010176.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700031087.mm.1 |
NM_010176.4 |
Refseq |