Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103182.2 | NP_001096652.2 | 2 - 3 | 282 | 108 | |
NR_120498.1 | 2 - 3 | 282 | 108 | |||
GenBank mRNA | AF190325.1 | - | 2 - 3 | 98 | 108 | |
AK052618.1 | - | 2 - 3 | 80 | 108 | ||
AK089989.1 | - | 2 - 3 | 285 | 106 | ||
BC088992.1 | - | 2 - 3 | 72 | 108 | ||
BC099695.1 | - | 2 - 3 | 66 | 108 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |