Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309048_m1
          See other D030028A08RIK GE Assays ›
          Gene Symbol:
          D030028A08Rik
          Gene Name:
          RIKEN cDNA D030028A08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 96944146 - 96965060 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309048_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D030028A08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003293.3 5 - 6 1331 139
          GenBank mRNA AK030811.1 - 5 - 6 1288 139
          AK040855.1 - 5 - 6 1280 139
          AK050871.1 - 5 - 6 1313 139
          AK140062.1 - 2 - 3 301 139
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY727057.1 - 2 - 3 301 Chr. 11 - 96960118 - 96963911 [+] on Build GRCm38 139

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream pyridoxine 5'-phosphate oxidase Pnpo AI415282 -16.8kb Anti-sense Chr. 11 - 96937816 - 96944019 on NCBI Build GRCm38
          Downstream trans-acting transcription factor 6 Sp6 1110025J03Rik,
          AA591031,
          AI592962,
          Epfn,
          Klf14
          49.0kb Sense Chr. 11 - 97013569 - 97024739 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001523.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961562 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 AK050871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 AK140062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 NR_003293.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 AK030811.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 AK050871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 AK140062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 NR_003293.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P148289 AK030811.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616084 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75819 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160297 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961562 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 AK050871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 AK140062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 NR_003293.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 AK030811.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 AK050871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 AK140062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 NR_003293.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P148289 AK030811.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160297 - -
          Whole Mouse Genome G4122A A_52_P148289 AK050871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148289 AK140062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148289 NR_003293.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148289 AK030811.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148289 AK050871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148289 AK140062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148289 NR_003293.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148289 AK030811.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160297 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100014010.mm.1 TC1100001523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100014010.mm.1 NR_003293.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014010.mm.1 NR_003293.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014011.mm.1 TC1100001523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100014011.mm.1 NR_003293.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014011.mm.1 NR_003293.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014012.mm.1 TC1100001523.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100014012.mm.1 NR_003293.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014012.mm.1 NR_003293.3 Refseq
          Murine Genome U74v2 Set 167329_PM_at - -
          Murine Genome U74v2 Set 167329_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 656797 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106770048 - -
          Illumina mouse-6 v1.0 expression beadchip 102810075 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214817 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2476005 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2582485 - -
          Illumina Sentrix mouse-6.v1 bead array 102810075 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102810075 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254136 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0