Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026817.3 | NP_081093.2 | 6 - 7 | 559 | 79 | |
GenBank mRNA | AK209433.1 | - | 1 - 2 | 38 | 79 | |
AK210352.1 | - | 1 - 2 | 38 | 79 | ||
BC030376.1 | - | 6 - 7 | 526 | 5374611 | 79 | |
BQ887131.1 | - | 6 - 7 | 551 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |