Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01307480_m1
          See other 4921509O07RIK GE Assays ›
          Gene Symbol:
          4921509O07Rik
          Gene Name:
          RIKEN cDNA 4921509O07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 113446288 - 113502844 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01307480_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4921509O07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045501.1 6 - 7 2059 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cDNA sequence BC067074 BC067074 -75.8kb Anti-sense Chr. 13 - 113292430 - 113370515 on NCBI Build GRCm38
          Downstream predicted gene,
          34471
          Gm34471 56.6kb Sense Chr. 13 - 113507828 - 113568698 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002694.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2353249 AK014856.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 327131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13258 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176903 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08002255 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02780920 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08002243 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176903 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08002281 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112474 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02780920 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176903 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08002255 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454017_PM_at - -
          Mouse Genome 430 2.0 Array 1454017_PM_at - -
          Mouse Genome 430 2.0 Array 1454017_at - -
          Affymetrix Mouse Clariom S Array PSR1300018244.mm.1 TC1300002694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018244.mm.1 NR_045501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018244.mm.1 NR_045501.1 Refseq
          Mouse Expression Set 430 1454017_PM_at - -
          Mouse Expression Set 430 1454017_at - -
          Affymetrix HT Mouse Genome 430B Array 1454017_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454017_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 639786 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0