Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01306487_m1
          See other SPN-PS GE Assays ›
          Gene Symbol:
          Spn-ps
          Gene Name:
          sialophorin, pseudogene
          Gene Aliases:
          Cd43-ps
          Chromosome Location:
          Chr.15: 90891396 - 90898476 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01306487_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spn-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033583.1 2 - 3 712 73
          GenBank mRNA AK007258.1 - 2 - 3 713 73
          BC048643.1 - 2 - 3 652 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002070.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325682 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131210 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131210 - -
          Whole Mouse Genome G4122A A_51_P469979 BC048643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469979 AK007258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469979 NR_033583.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112650 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12011084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12096200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12011079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12376239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12445557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11919604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12376240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30432497 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432479_PM_at - -
          Mouse Genome 430 2.0 Array 1432479_at - -
          Mouse Genome 430 2.0 Array 1432479_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500016703.mm.1 TC1500002070.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016703.mm.1 NR_033583.1 Refseq
          Mouse Expression Set 430 1432479_at - -
          Mouse Expression Set 430 1432479_PM_at - -
          Mouse Genome 430A 2.0 Array 1432479_PM_at - -
          Mouse Genome 430A 2.0 Array 1432479_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 777053 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0