Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01303067_m1
          See other WSB1 GE Assays ›
          Gene Symbol:
          Wsb1
          Gene Name:
          WD repeat and SOCS box-containing 1
          Gene Aliases:
          1110056B13Rik, 2700038M07Rik
          Chromosome Location:
          Chr.11: 79239382 - 79254700 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01303067_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wsb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019653.3 NP_062627.3 1 - 2 338 78
          GenBank mRNA AK019206.2 - 1 - 2 315 78
          AK145974.1 - 1 - 2 317 78
          AK146752.1 - 1 - 2 313 78
          AK150394.1 - 1 - 2 313 78
          AK150600.1 - 1 - 2 313 78
          AK152545.1 - 1 - 2 316 78
          AK159905.1 - 1 - 2 313 78
          AK167174.1 - 1 - 2 315 78
          AK167394.1 - 1 - 2 316 78
          AK167942.1 - 1 - 2 313 78
          AK168468.1 - 1 - 2 316 78
          AK168565.1 - 1 - 2 316 78
          AK169058.1 - 1 - 2 313 78
          BC053391.1 - 1 - 2 89 30073116 78
          BY097910.1 - 1 - 2 339 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003318.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK145974.1, AK146752.1, AK150394.1, AK150600.1, AK152545.1, AK159905.1, AK167174.1, AK167394.1, AK167942.1, AK169058.1, BC053391.1, NM_019653.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wsb1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          intracellular signal transduction
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100030570.mm.1 TC1100003318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030570.mm.1 NM_019653.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030570.mm.1 NM_019653.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030572.mm.1 TC1100003318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030572.mm.1 NM_019653.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030572.mm.1 NM_019653.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030571.mm.1 TC1100003318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030571.mm.1 NM_019653.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030571.mm.1 NM_019653.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030573.mm.1 TC1100003318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030573.mm.1 NM_019653.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030573.mm.1 NM_019653.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 702705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0