Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01302571_g1
          See other CCDC154 GE Assays ›
          Gene Symbol:
          Ccdc154
          Gene Name:
          coiled-coil domain containing 154
          Gene Aliases:
          Ccdc157, Gm317
          Chromosome Location:
          Chr.17: 25162168 - 25171913 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01302571_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc154
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001079929.2 NP_001073398.2 15 - 16 2065 72
          GenBank mRNA AK133084.1 - 16 - 17 2182 72
          BB017594.2 - 4 - 5 326 72
          JN935900.1 - 14 - 15 1684 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000363.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994144 NM_001079929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994144 AK133084.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08741731 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08741708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13326286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13454036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13171128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13076961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13076966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13432317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10442719 - -
          Affymetrix Mouse Clariom S Array PSR1700003005.mm.1 TC1700000363.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003005.mm.1 NM_001079929.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003004.mm.1 TC1700000363.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003004.mm.1 NM_001079929.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003002.mm.1 TC1700000363.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003002.mm.1 NM_001079929.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003001.mm.1 TC1700000363.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003001.mm.1 NM_001079929.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 333036 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250498 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0