Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01302472_m1
          See other PRSS32 GE Assays ›
          Gene Symbol:
          Prss32
          Gene Name:
          protease, serine 32
          Gene Aliases:
          2010001P08Rik, mT5, tryptase-5
          Chromosome Location:
          Chr.17: 23853772 - 23859776 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01302472_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss32
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027220.2 NP_081496.2 4 - 5 763 87
          GenBank mRNA AY266139.1 - 4 - 5 750 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000325.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss32

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          serine-type endopeptidase activity
          serine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966862 BC024903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966862 NM_027220.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966862 AY266139.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90645 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150640 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966862 BC024903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966862 NM_027220.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966862 AY266139.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150640 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08737829 - -
          Whole Mouse Genome G4122A A_52_P344601 BC024903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P344601 NM_027220.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P344601 AK008023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P344601 AY266139.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13539838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13539833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13179608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13212160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13212152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13330793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13539833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094034 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442358 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442362 - -
          Affymetrix HT MG-430 PM Array Plate 1454267_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1424689_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442357 - -
          Mouse Genome 430 2.0 Array 1454267_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424689_PM_at - -
          Mouse Genome 430 2.0 Array 1424689_at - -
          Mouse Genome 430 2.0 Array 1454267_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69814_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69814_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700002383.mm.1 TC1700000325.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002383.mm.1 NM_027220.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002383.mm.1 NM_027220.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002384.mm.1 TC1700000325.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002384.mm.1 NM_027220.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002384.mm.1 NM_027220.2 Refseq
          Mouse Expression Set 430 1454267_PM_a_at - -
          Mouse Expression Set 430 1424689_PM_at - -
          Mouse Expression Set 430 1454267_a_at - -
          Mouse Expression Set 430 1424689_at - -
          Murine Genome U74v2 Set 104185_PM_at - -
          Murine Genome U74v2 Set 162205_PM_f_at - -
          Murine Genome U74v2 Set 162205_f_at - -
          Murine Genome U74v2 Set 104185_at - -
          Mouse Genome 430A 2.0 Array 1424689_at - -
          Mouse Genome 430A 2.0 Array 1454267_a_at - -
          Mouse Genome 430A 2.0 Array 1424689_PM_at - -
          Mouse Genome 430A 2.0 Array 1454267_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424689_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1454267_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424689_at - -
          Affymetrix HT Mouse Genome 430A Array 1454267_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5670707 - -
          Illumina mouse-6 v1.0 expression beadchip 106770093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237586 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244093 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237586 - -
          Illumina Sentrix mouse-6.v1 bead array 106770093 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106770093 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244093 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0