Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01300919_m1
          See other PLEKHD1OS GE Assays ›
          Gene Symbol:
          Plekhd1os
          Gene Name:
          pleckstrin homology domain containing, family D (with coiled-coil domains) member 1, opposite strand
          Gene Aliases:
          0610009B14Rik
          Chromosome Location:
          Chr.12: 80686375 - 80693952 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01300919_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plekhd1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037995.1 1 - 2 309 104
          GenBank mRNA AK002366.1 - 1 - 2 309 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002005.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK002366.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321930 AK002366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321930 NR_037995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321930 AK002366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321930 NR_037995.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154584 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321930 AK002366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321930 NR_037995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321930 AK002366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321930 NR_037995.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154584 - -
          Whole Mouse Genome G4122A A_51_P321930 AK002366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321930 NR_037995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321930 AK002366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321930 NR_037995.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09796389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09763675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10104728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09914561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09763683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09914569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28577062 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431446_PM_at - -
          Mouse Genome 430 2.0 Array 1431446_PM_at - -
          Mouse Genome 430 2.0 Array 1431446_at - -
          Affymetrix Mouse Clariom S Array PSR1200013796.mm.1 TC1200002005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013796.mm.1 NR_037995.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013796.mm.1 NR_037995.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013797.mm.1 TC1200002005.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200013797.mm.1 NR_037995.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200013797.mm.1 NR_037995.1 Refseq
          Mouse Expression Set 430 1431446_PM_at - -
          Mouse Expression Set 430 1431446_at - -
          Murine Genome U74v2 Set 165906_PM_at - -
          Murine Genome U74v2 Set 165906_at - -
          Affymetrix HT Mouse Genome 430B Array 1431446_at - -
          Affymetrix HT Mouse Genome 430B Array 1431446_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 340871 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0