Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292035_m1
          See other 1810007C17RIK GE Assays ›
          Gene Symbol:
          1810007C17Rik
          Gene Name:
          RIKEN cDNA 1810007C17 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 49476993 - 49480862 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292035_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810007C17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045472.1 2 - 3 472 153
          GenBank mRNA AK007358.1 - 2 - 3 476 153

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 3110039M20 gene 3110039M20Rik -69.6kb Anti-sense Chr. 12 - 49389652 - 49407346 on NCBI Build GRCm38
          Downstream protein kinase D1 Prkd1 PKD,
          PKD1,
          Pkcm,
          Prkcm
          863.1kb Sense Chr. 12 - 50341226 - 50649248 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908973 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131189 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10326351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10326356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10326347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10144427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10326338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28420498 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 169716_at - -
          Murine Genome U74v2 Set 169716_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0