Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01292015_g1
          See other MCCC1OS GE Assays ›
          Gene Symbol:
          Mccc1os
          Gene Name:
          methylcrotonoyl-Coenzyme A carboxylase 1 (alpha), opposite strand
          Gene Aliases:
          A330050B17Rik
          Chromosome Location:
          Chr.3: 35999913 - 36008828 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01292015_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mccc1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029456.1 2 - 3 934 71
          GenBank mRNA AK032105.1 - 2 - 3 936 71
          AK036093.1 - 413 71
          AK138522.1 - 2 - 3 936 71

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2296566 AK138522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2296566 NR_029456.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78325 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2296566 AK138522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2296566 NR_029456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500247 AK039056.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500247 AK138522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500247 AK032105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500247 NR_029456.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10491525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491523 - -
          Affymetrix Mouse Gene 1.1 ST Array 10497794 - -
          Affymetrix Mouse Gene 1.1 ST Array 10491522 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 828248 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104810471 - -
          Illumina mouse-6 v1.0 expression beadchip 104810471 - -
          Illumina mouse-6 v1.0 expression beadchip 103520195 - -
          Illumina mouse-6 v1.0 expression beadchip 102570594 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243425 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238437 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2554920 - -
          Illumina Sentrix mouse-6.v1 bead array 104810471 - -
          Illumina Sentrix mouse-6.v1 bead array 102570594 - -
          Illumina Sentrix mouse-6.v1 bead array 103520195 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102570594 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104810471 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103520195 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2554920 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243425 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238437 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7d7666c9cf-ndzlw:80/100.66.129.171:80.
          git-commit: 173eb311d069d8bb3c8868db8478582b9fe5b4f6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0