Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291695_m1
          See other GM19990 GE Assays ›
          Gene Symbol:
          Gm19990
          Gene Name:
          predicted gene, 19990
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 56253243 - 56266381 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291695_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm19990
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045048.1 3 - 4 284 68
          GenBank mRNA AK053248.1 - 3 - 4 285 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB664650.1 - 3 - 4 282 Chr. 12 - 56253245 - 56265813 [+] on Build GRCm38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream breast cancer metastasis-suppressor 1-like Brms1l 0710008O11Rik,
          AI159718,
          BRMS1,
          D12Ertd407e
          -394.2kb Sense Chr. 12 - 55836243 - 55869736 on NCBI Build GRCm38
          Downstream MAP3K12 binding inhibitory protein 1 Mbip 4933408E06Rik 61.9kb Anti-sense Chr. 12 - 56328306 - 56345894 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000475.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685770 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09907128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10026374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10285894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28455683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10248856 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200003283.mm.1 TC1200000475.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003283.mm.1 NR_045048.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003283.mm.1 NR_045048.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003284.mm.1 TC1200000475.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003284.mm.1 NR_045048.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003284.mm.1 NR_045048.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 627098 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0