Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281912_m1
          See other NOS1AP GE Assays ›
          Gene Symbol:
          Nos1ap
          Gene Name:
          nitric oxide synthase 1 (neuronal) adaptor protein
          Gene Aliases:
          6330408P19Rik, Capon
          Chromosome Location:
          Chr.1: 170317414 - 170589849 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281912_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nos1ap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109985.1 NP_001103455.1 3 - 4 755 92
          NM_027528.2 NP_081804.1 3 - 4 572 92
          GenBank mRNA AK045124.1 - 3 - 4 709 92
          AK053892.1 - 3 - 4 756 92
          AK138471.1 - 3 - 4 573 92
          AK162919.1 - 3 - 4 573 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003887.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of neuron death
          regulation of high voltage-gated calcium channel activity
          positive regulation of peptidyl-cysteine S-nitrosylation
          protease binding
          PDZ domain binding
          nitric-oxide synthase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100028723.mm.1 TC0100003887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028723.mm.1 NM_001109985.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028723.mm.1 NM_001109985.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028723.mm.1 NM_027528.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028723.mm.1 NM_027528.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028725.mm.1 TC0100003887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028725.mm.1 NM_001109985.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028725.mm.1 NM_001109985.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028725.mm.1 NM_027528.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028725.mm.1 NM_027528.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028729.mm.1 TC0100003887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028729.mm.1 NM_027528.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028729.mm.1 NM_027528.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028728.mm.1 TC0100003887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028728.mm.1 NM_001109985.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028728.mm.1 NM_001109985.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028728.mm.1 NM_027528.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028728.mm.1 NM_027528.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028730.mm.1 TC0100003887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028730.mm.1 NM_001109985.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028730.mm.1 NM_001109985.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028731.mm.1 TC0100003887.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100028731.mm.1 NM_001109985.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100028731.mm.1 NM_001109985.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 491275 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0