Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281515_g1
          See other DNPH1 GE Assays ›
          Gene Symbol:
          Dnph1
          Gene Name:
          2'-deoxynucleoside 5'-phosphate N-hydrolase 1
          Gene Aliases:
          C76683, Rcl
          Chromosome Location:
          Chr.17: 46496789 - 46499618 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281515_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnph1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207161.3 NP_997044.2 3 - 4 364 66
          GenBank mRNA AK146723.1 - 3 - 4 365 66
          AK151589.1 - 3 - 4 348 66
          BC048355.1 - 3 - 4 353 5388297 66
          BC082791.1 - 3 - 4 390 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000896.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnph1

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          nucleotide metabolic process
          deoxyribonucleoside monophosphate catabolic process
          positive regulation of cell growth
          purine-specific mismatch base pair DNA N-glycosylase activity
          uracil DNA N-glycosylase activity
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds
          single-strand selective uracil DNA N-glycosylase activity
          DNA N-glycosylase activity
          protein homodimerization activity
          DNA-3-methylbase glycosylase activity
          nucleoside deoxyribosyltransferase activity
          nicotinamide riboside hydrolase activity
          nicotinic acid riboside hydrolase activity
          deoxyribonucleoside 5'-monophosphate N-glycosidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207194 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207195 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207191 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207192 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 AK151589.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 BC082791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 NM_207161.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 AK146723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 AK151589.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 BC082791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 NM_207161.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P596755 AK146723.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892787 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107526 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 AK151589.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 BC082791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 NM_207161.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 AK146723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 AK151589.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 BC082791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 NM_207161.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P596755 AK146723.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31207243 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107526 - -
          Whole Mouse Genome G4122A A_52_P596755 AK151589.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P596755 BC082791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P596755 NM_207161.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P596755 AK146723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P596755 AK151589.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P596755 BC082791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P596755 NM_207161.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P596755 AK146723.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101186 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107526 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13090515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13090512 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13090511 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13092799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207195 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207191 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207194 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31207192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31207200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13382698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13062185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13349337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13198286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13062184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13212242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13445282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13450212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13090515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31207213 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445559 - -
          Affymetrix HT MG-430 PM Array Plate 1460713_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445558 - -
          Mouse Genome 430 2.0 Array 1460713_at - -
          Mouse Genome 430 2.0 Array 1460713_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381101_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381101_at - -
          Affymetrix Mouse Clariom S Array PSR1700008539.mm.1 TC1700000896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008539.mm.1 NM_207161.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008539.mm.1 NM_207161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008540.mm.1 TC1700000896.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008540.mm.1 NM_207161.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008540.mm.1 NM_207161.3 Refseq
          Mouse Expression Set 430 1460713_at - -
          Mouse Expression Set 430 1460713_PM_at - -
          Murine Genome U74v2 Set 97179_at - -
          Murine Genome U74v2 Set 97179_PM_at - -
          Mouse Genome 430A 2.0 Array 1460713_at - -
          Mouse Genome 430A 2.0 Array 1460713_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460713_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460713_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3120601 - -
          Illumina mouse-6 v1.0 expression beadchip 3120601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2676290 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2676290 - -
          mouse-6 v1.1 (Illumina) scl0381101.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0381101.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120601 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2676294 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2676294 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0