Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01281279_m1
          See other ADGRF2 GE Assays ›
          Gene Symbol:
          Adgrf2
          Gene Name:
          adhesion G protein-coupled receptor F2
          Gene Aliases:
          Gpr111, PGR20
          Chromosome Location:
          Chr.17: 42694028 - 42749145 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01281279_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adgrf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033493.2 NP_001028665.2 5 - 6 800 108
          GenBank mRNA AK142836.1 - 5 - 6 802 108
          BC141353.1 - 5 - 6 703 9055965 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002117.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011595 NM_001033493.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011595 AK142836.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643186 BC116724.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643186 NM_008458.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783078 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131427 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137521 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011595 NM_001033493.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011595 AK142836.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03318183 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137521 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03318200 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03318191 - -
          Whole Mouse Genome G4122A A_52_P615630 BC141353.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615630 NM_001033493.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615630 AK142836.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137521 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03318200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31185685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13444927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13129809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13468768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13468732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13462724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13200823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13378803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13175189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13187310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31185757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451000 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450998 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450996 - -
          Affymetrix Mouse Clariom S Array PSR1700019813.mm.1 TC1700002117.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019813.mm.1 NM_001033493.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019813.mm.1 NM_001033493.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019814.mm.1 TC1700002117.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019814.mm.1 NM_001033493.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019814.mm.1 NM_001033493.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161164 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161164 - -
          mouse-6 v1.1 (Illumina) GI_85702032-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161164 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-88d55b78f-jn859:80/100.66.134.75:80.
          git-commit: 0e87aa2f016cbc64993b9746d11f1634ddfab556
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0