Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700008139.mm.1 |
TC1700000850.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008139.mm.1 |
NM_178652.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008139.mm.1 |
NM_178652.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008140.mm.1 |
TC1700000850.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008140.mm.1 |
NM_178652.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008140.mm.1 |
NM_178652.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008143.mm.1 |
TC1700000850.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008143.mm.1 |
NM_178652.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008143.mm.1 |
NM_178652.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008141.mm.1 |
TC1700000850.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700008141.mm.1 |
NM_178652.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700008141.mm.1 |
NM_178652.2 |
Refseq |