Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01261944_g1
          See other 1110008L16RIK GE Assays ›
          Gene Symbol:
          1110008L16Rik
          Gene Name:
          RIKEN cDNA 1110008L16 gene
          Gene Aliases:
          Kiaa0391, Mrpp3, mKIAA0391
          Chromosome Location:
          Chr.12: 55302637 - 55382491 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01261944_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1110008L16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025373.1 NP_079649.1 7 3063 60
          GenBank mRNA AK003589.1 - 7 993 60
          AK008749.1 - 3 489 60
          AK031144.1 - 7 2441 60
          AK045434.1 - 7 2431 60
          AK151976.1 - 7 3063 60
          AK152028.1 - 7 3063 60
          AK172942.1 - 7 867 60
          BC034876.1 - 7 1915 1512335 60
          BC037485.1 - 7 1419 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000464.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA processing
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112609 AK152028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112609 BC034876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112609 AK045434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112609 AK151976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112609 AK031144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 BC037485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 AK152028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 BC034876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 AK045434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 AK151976.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 BC037485.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 AK152028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 BC034876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 AK045434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141953 AK151976.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119540 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 656100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44237 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163479 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99083 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136716 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162415 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112609 AK152028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112609 BC034876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112609 AK045434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112609 AK151976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112609 AK031144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 BC037485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 AK152028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 BC034876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 AK045434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 AK151976.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 BC037485.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 AK152028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 BC034876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 AK045434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141953 AK151976.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 BC037485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 AK152028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 BC034876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 AK045434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 AK151976.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 BC037485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 AK152028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 BC034876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 AK045434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141953 AK151976.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10337115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09814208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09841322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10419343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09973947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09844157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10055634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10130580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10028163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10067492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09773909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09815030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09801540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10019955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10028156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09815033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09864487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09773907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10331875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10384218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10224936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09933479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09801541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10337103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09841317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09884794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09815035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09960416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10318404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09837601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10288788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28451520 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10395808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395809 - -
          Affymetrix HT MG-430 PM Array Plate 1428775_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10395807 - -
          Mouse Genome 430 2.0 Array 1428775_PM_at - -
          Mouse Genome 430 2.0 Array 1428775_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66132_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66132_at - -
          Affymetrix Mouse Clariom S Array PSR1200003220.mm.1 TC1200000464.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003220.mm.1 NM_025373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003220.mm.1 NM_025373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003221.mm.1 TC1200000464.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003221.mm.1 NM_025373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003221.mm.1 NM_025373.1 Refseq
          Mouse Expression Set 430 1428775_PM_at - -
          Mouse Expression Set 430 1428775_at - -
          Affymetrix HT Mouse Genome 430B Array 1428775_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428775_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100070711 - -
          Illumina mouse-6 v1.0 expression beadchip 100070711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2668013 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2576227 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2668013 - -
          Illumina Sentrix mouse-6.v1 bead array 100070711 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100070711 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2576227 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5b7c5955bc-w9n6d:80/100.66.134.79:80.
          git-commit: d64a76636f5352962c24cd1b66db12392f7c7b6b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0