Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01261182_s1
          See other HIST2H3C2 GE Assays ›
          Gene Symbol:
          Hist2h3c2
          Gene Name:
          histone cluster 2, H3c2
          Gene Aliases:
          H3-614, H3.2-614, Hist2h3c2-ps, Hist2h3ca1, M32461
          Chromosome Location:
          Chr.3: 96238479 - 96239127 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01261182_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist2h3c2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA CO040940.1 - 1 80

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2142251 NM_054045.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2142251 BC015270.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2142251 NM_054045.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2142251 BC015270.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348328 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04997256 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04997256 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02709729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02653277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03172954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02653272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02966250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348322 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494400 - -
          Affymetrix HT MG-430 PM Array Plate 1422155_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10500327 - -
          Affymetrix Mouse Gene 1.1 ST Array 10494405 - -
          Affymetrix Mouse Gene 1.1 ST Array 10494402 - -
          Affymetrix Mouse Gene 1.1 ST Array 10494395 - -
          Mouse Genome 430 2.0 Array 1422155_at - -
          Mouse Genome 430 2.0 Array 1422155_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 97114_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 97114_PM_at - -
          Mouse Expression Set 430 1418367_PM_x_at - -
          Mouse Expression Set 430 1460314_PM_s_at - -
          Mouse Expression Set 430 1418366_PM_at - -
          Mouse Expression Set 430 1453573_at - -
          Mouse Expression Set 430 1427864_PM_at - -
          Mouse Expression Set 430 1418366_at - -
          Mouse Expression Set 430 1418367_x_at - -
          Mouse Expression Set 430 1453573_PM_at - -
          Mouse Expression Set 430 1460314_s_at - -
          Mouse Expression Set 430 1422155_at - -
          Mouse Expression Set 430 1427864_at - -
          Mouse Expression Set 430 1422155_PM_at - -
          Mouse Expression Set 430 1442051_PM_at - -
          Mouse Expression Set 430 1442051_at - -
          Murine Genome U74v2 Set 93068_r_at - -
          Murine Genome U74v2 Set 94756_at - -
          Murine Genome U74v2 Set 93067_f_at - -
          Murine Genome U74v2 Set 93022_PM_i_at - -
          Murine Genome U74v2 Set 93022_i_at - -
          Murine Genome U74v2 Set 96416_f_at - -
          Murine Genome U74v2 Set 93068_PM_r_at - -
          Murine Genome U74v2 Set 93067_PM_f_at - -
          Murine Genome U74v2 Set 93023_PM_f_at - -
          Murine Genome U74v2 Set 94756_PM_at - -
          Murine Genome U74v2 Set 96416_PM_f_at - -
          Murine Genome U74v2 Set 93023_f_at - -
          Mouse Genome 430A 2.0 Array 1460314_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1453573_at - -
          Mouse Genome 430A 2.0 Array 1418367_x_at - -
          Mouse Genome 430A 2.0 Array 1422155_at - -
          Mouse Genome 430A 2.0 Array 1453573_PM_at - -
          Mouse Genome 430A 2.0 Array 1427864_at - -
          Mouse Genome 430A 2.0 Array 1418366_PM_at - -
          Mouse Genome 430A 2.0 Array 1422155_PM_at - -
          Mouse Genome 430A 2.0 Array 1418367_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1460314_s_at - -
          Mouse Genome 430A 2.0 Array 1418366_at - -
          Mouse Genome 430A 2.0 Array 1427864_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422155_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422155_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3990670 - -
          Illumina mouse-6 v1.0 expression beadchip 3990670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244200 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724358 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244200 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724358 - -
          Illumina Sentrix mouse-6.v1 bead array 3990670 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21816.1.35_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990670 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5f68d85665-v9rd4:80/100.66.130.250:80.
          git-commit: a5c360236fb87e3c46c4dd6f38a1a1c611be0521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0