Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01251006_m1
          See other CES3A GE Assays ›
          Gene Symbol:
          Ces3a
          Gene Name:
          carboxylesterase 3A
          Gene Aliases:
          Ces3, Ces31, Es31
          Chromosome Location:
          Chr.8: 105048577 - 105061928 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01251006_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164681.1 NP_001158153.1 8 - 9 1025 65
          NM_198672.1 NP_941074.1 9 - 10 1166 65
          GenBank mRNA AK138932.1 - 9 - 10 1165 65
          AK149499.1 - 5 - 6 586 65
          BC035526.1 - 8 - 9 1021 65
          BC048380.1 - 9 - 10 1147 65
          BC057187.1 - 9 - 10 1151 5098677 65
          BC057188.1 - 9 - 10 1151 5100121 65
          BC061004.1 - 8 - 9 1023 30315941 65
          L11333.1 - 9 - 10 1141 65
          S64130.1 - 9 - 10 1142 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001200.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          hydrolase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061219 S64130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061219 BC057188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061219 BC061004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061219 L11333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2061219 BC035526.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101735 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061219 S64130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061219 BC057188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061219 BC061004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061219 L11333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2061219 BC035526.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06625495 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101735 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07392663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06932180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07229432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06989708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26131384 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574602 - -
          Affymetrix Mouse Gene 1.1 ST Array 10574598 - -
          Affymetrix Mouse Clariom S Array PSR0800009393.mm.1 TC0800001200.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009393.mm.1 NM_198672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009393.mm.1 NM_198672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009395.mm.1 TC0800001200.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009395.mm.1 NM_001164681.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009395.mm.1 NM_001164681.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009395.mm.1 NM_198672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009395.mm.1 NM_198672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009397.mm.1 TC0800001200.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009397.mm.1 NM_001164681.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009397.mm.1 NM_001164681.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009397.mm.1 NM_198672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009397.mm.1 NM_198672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009396.mm.1 TC0800001200.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009396.mm.1 NM_001164681.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009396.mm.1 NM_001164681.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009396.mm.1 NM_198672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009396.mm.1 NM_198672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009398.mm.1 TC0800001200.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009398.mm.1 NM_001164681.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009398.mm.1 NM_001164681.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009398.mm.1 NM_198672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009398.mm.1 NM_198672.1 Refseq
          Mouse Expression Set 430 1451600_PM_s_at - -
          Mouse Expression Set 430 1451600_s_at - -
          Murine Genome U74v2 Set 99941_PM_at - -
          Murine Genome U74v2 Set 99941_at - -
          Mouse Genome 430A 2.0 Array 1451600_s_at - -
          Mouse Genome 430A 2.0 Array 1451600_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240872 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240872 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0